missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GMEB1 Full-length ORF (NP_077808.1, 1 a.a. - 563 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00010691-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16125582

  • 574.00 CHF / 25 Mikrogramm
Verfügbar ab: 17-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene is a member of KDWK gene family. The product of this gene associates with GMEB2 protein, and the complex is essential for parvovirus DNA replication. Study of rat homolog implicates the role of this gene in modulation of transactivation by the glucocorticoid receptor bound to glucocorticoid response elements. Two alternative spliced transcript variants encoding different isoforms exist. [provided by RefSeq]

Sequence: MANAEVSVPVGDVVVVPTEGNEGENPEDTKTQVILQLQPVQQGIYEAGSENNTAVVAVETHTIHKIEEGIDTGTIEANEDMEIAYPITCGESKAILLWKKFVCPGINVKCVKFNDQLISPKHFVHLAGKSTLKDWKRAIRLGGIMLRKMMDSGQIDFYQHDKVCSNTCRSTKFDLLISSARAPVPGQQTSVVQTPTSADGSITQIAISEESMEEAGLEWNSALTAAVTMATEEGVKKDSEEISEDTLMFWKGIADVGLMEEVVCNIQKEIEELLRGVQQRLIQAPFQVTDAAVLNNVAHTFGLMDTVKKVLDNRRNQVEQGEEQFLYTLTDLERQLEEQKKQGQDHRLKSQTVQNVVLMPVSTPKPPKRPRLQRPASTTVLSPSPPVQQPQFTVISPITITPVGQSFSMGNIPVATLSQGSSPVTVHTLPSGPQLFRYATVVSSAKSSSPDTVTIHPSSSLALLSSTAMQDGSTLGNMTTMVSPVELVAMESGLTSAIQAVESTSEDGQTIIEIDPAPDPEAEDTEGKAVILETELRTEEKVVAEMEEHQHQVHNVEIVVLED
Spezifikation

Spezifikation

NP_077808.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
P96PIF/PIF96
GMEB1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10691
GMEB1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MANAEVSVPVGDVVVVPTEGNEGENPEDTKTQVILQLQPVQQGIYEAGSENNTAVVAVETHTIHKIEEGIDTGTIEANEDMEIAYPITCGESKAILLWKKFVCPGINVKCVKFNDQLISPKHFVHLAGKSTLKDWKRAIRLGGIMLRKMMDSGQIDFYQHDKVCSNTCRSTKFDLLISSARAPVPGQQTSVVQTPTSADGSITQIAISEESMEEAGLEWNSALTAAVTMATEEGVKKDSEEISEDTLMFWKGIADVGLMEEVVCNIQKEIEELLRGVQQRLIQAPFQVTDAAVLNNVAHTFGLMDTVKKVLDNRRNQVEQGEEQFLYTLTDLERQLEEQKKQGQDHRLKSQTVQNVVLMPVSTPKPPKRPRLQRPASTTVLSPSPPVQQPQFTVISPITITPVGQSFSMGNIPVATLSQGSSPVTVHTLPSGPQLFRYATVVSSAKSSSPDTVTIHPSSSLALLSSTAMQDGSTLGNMTTMVSPVELVAMESGLTSAIQAVESTSEDGQTIIEIDPAPDPEAEDTEGKAVILETELRTEEKVVAEMEEHQHQVHNVEIVVLED
RUO
GMEB1
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote