Learn More
Human GLI4 knockout HEK-293T cell lysate, 1 Kit
Product Code. 32156078 Shop All Abcam Products
Product Code. 32156078 Supplier Abcam Supplier No. ab2632161Kit
 Restricted Item
Verfügbar ab: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Human GLI4 knockout HEK-293T cell/tissue lysate.
  • Species: Human
  • Tissue: GLI4 knockout HEK-293T