missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GFR alpha-like Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Marke: R&D Systems MAB9697-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
GFR alpha-like Monoclonal specifically detects GFR alpha-like in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.
Spezifikation
| GFR alpha-like | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry, Blocking Assay, CyTOF | |
| 2345C | |
| Flow Cytometry 0.25 μg/106 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| Q6UXV0 | |
| Human embryonic kidney cell line HEK293-derived recombinant human GFR alpha-like, Ser19-Glu351, Accession # Q6UXV0 | |
| 100 μg | |
| Detects human GFR alpha-like in direct ELISAs. | |
| Human | |
| Supernatant |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur