missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human GAPDH / G3PDH Protein, His Tag

Marke:  ACROBiosystems GAH-H5145-1MG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15826817

  • 1128.43 CHF / 1 Milligramm


Verfügbar ab
26-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) is also known as Peptidyl-cysteine S-nitrosylase GAPDH, which is an enzyme that catalyzes the sixth step of glycolysis and thus serves to break down glucose for energy and carbon molecules. GAPDH has both glyceraldehyde-3-phosphate dehydrogenase and nitrosylase activities, thereby playing a role in glycolysis and nuclear functions, respectively. GAPDH can participate in nuclear events including transcription, RNA transport, DNA replication and apoptosis. Also, GAPDH can modulate the organization and assembly of the cytoskeleton. Furthermore, GAPDH is a component of the GAIT (gamma interferon-activated inhibitor of translation) complex which mediates interferon-gamma-induced transcript-selective translation inhibition in inflammation processes.

This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.9 kDa. The protein migrates as 37-38 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation

Spezifikation

AAH01601
E.coli
Human
36.9 kDa
1 mg
Gly 2 - Glu 335
GAPDH,GAPD,CDABP0047,OK/SW-cl0.12
Unconjugated
50 mM Tris, 150 mM NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
Other Recombinant Proteins
Powder
>95% as determined by SDS-PAGE
Human GAPDH, His Tag (GAH-H5145) is expressed from E.coli cells. It contains AA Gly 2 - Glu 335 (Accession # AAH01601).
-20°C
GAPDH
His Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation