Learn More
ACROBiosystems Human Galectin-3 Protein, His Tag
Marke: ACROBiosystems GA3-H5129-200UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Galectin-3 (Gal-3) is also known as LGALS3, 35 kDa lectin, Carbohydrate-binding protein 35 (CBP 35), Galactose-specific lectin 3, Galactoside-binding protein (GALBP), IgE-binding protein, Laminin-binding protein, Mac-2 antigen, Lectin L-29. Galectin-3 is a member of the lectin family. LGALS3 / Galectin-3 is expressed in the nucleus, cytoplasm, mitochondrion, cell surface, and extracellular space. LGALS3 / Galectin-3 has been shown to be involved in the following biological processes: cell adhesion, cell activation and chemoattraction, cell growth and differentiation, cell cycle, and apoptosis.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.0 kDa. The protein migrates as 32 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
NP_002297 | |
200 μg | |
Met 1 - Ile 250 | |
Galectin-3,LGALS3,MAC2,Gal-3,35 kDa lectin,CBP 35,GALBP,IgE-binding protein,L-31,Laminin-binding protein,Lectin L-29,Mac-2 antigen | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
27.0 kDa | |
Human Galectin-3, His Tag (GA3-H5129) is expressed from E.coli cells. It contains AA Met 1 - Ile 250 (Accession # NP_002297). | |
-20°C | |
Galectin-3 | |
Human | |
50 mM HEPES, 150 mM NaCl, 5 mM EDTA, 2 mM TCEP, pH7.4 | |
E.coli | |
Bio-Markers and CD Antigens |