missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FNDC5 ELISA Kit
For the quantitative detection of Human FNDC5
Marke: Invitrogen™ EEL081
Additional Details : Gewicht : 0.40000kg
Beschreibung
Fibronectin type III domain-containing protein 5, the precursor of irisin, is a protein that is encoded by the FNDC5 gene. This gene encodes a secreted protein that is released from muscle cells during exercise. The encoded protein may participate in the development of brown fat. Translation of the precursor protein initiates at a non-AUG start codon at a position that is conserved as an AUG start codon in other organisms. Alternative splicing results in multiple transcript variants.Spezifikation
Q8NAU1 | |
9.38 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
FNDC5 | |
<10% | |
HRP | |
RUO | |
Refer to product documentation for component specific storage temperature. | |
1 hr 20 min |
15.63 to 1000 pg/mL | |
HRP | |
Serum | |
252995 | |
<10% | |
Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer | |
96 Tests | |
100 µL | |
Human | |
3 hr 30 min |