missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FNDC1 Partial ORF (XP_027658.3, 86 a.a. - 184 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00084624-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: VPSRLPPRSAATVSPVAGTHPWPQYTTRAPPGHFSTTPMLSLRQRMMHARFRNPLSRQPARPSYRQGYNGRPNVEGKVLPGSNGKPNGQRIINGPQGTKSpezifikation
XP_027658.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VPSRLPPRSAATVSPVAGTHPWPQYTTRAPPGHFSTTPMLSLRQRMMHARFRNPLSRQPARPSYRQGYNGRPNVEGKVLPGSNGKPNGQRIINGPQGTK | |
RUO | |
FNDC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84624 | |
FNDC1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AGS8/FNDC2/KIAA1866/MEL4B3/bA243O10.1/dJ322A24.1 | |
FNDC1 | |
Recombinant | |
wheat germ expression system |