missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FN1 native Protein from human plasma

Artikelnummer. 16038096
Click to view available options
Menge:
1 mg
Packungsgröße:
1 Milligramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 16038096

Marke: Abnova™ P4955.1mg

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Used for SDS-PAGE

This gene encodes fibronectin, a glycoprotein present in a soluble dimeric form in plasma, and in a dimeric or multimeric form at the cell surface and in extracellular matrix. Fibronectin is involved in cell adhesion and migration processes including embryogenesis, wound healing, blood coagulation, host defense, and metastasis. The gene has three regions subject to alternative splicing, with the potential to produce 20 different transcript variants. However, the full-length nature of some variants has not been determined. [provided by RefSeq]

Spezifikation

Konzentration 22.0 mg/mL
Zur Verwendung mit (Anwendung) SDS-PAGE
Zusammensetzung In 0.1M Tris-HCl, 0.15M NaCl, pH 7.4
Gen-ID (Entrez) 2335
Molekulargewicht 220kDa
Name FN1 (Human) Native Protein
Vorbereitungsmethode Native protein purified from human plasma
Reinigungsverfahren Column chromatography
Menge 1 mg
Lagerungsbedingungen Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Kennzeichnung RUO
Gen-Alias CIG/DKFZp686F10164/DKFZp686H0342/DKFZp686I1370/DKFZp686O13149/ED-B/FINC/FN/FNZ/GFND/GFND2/LETS/MSF
Gebräuchliche Bezeichnung FN1
Gensymbol FN1
Spezies Human
Proteinmarkierung None
Expressionssystem Native protein purified from human plasma
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.