missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human Flt-3 Ligand Protein, His Tag

207.26 CHF - 3224.10 CHF

Spezifikation

Quelle Human Flt-3 Ligand, His Tag (FLL-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Pro 185 (Accession # P49771-1).
Gebräuchliche Bezeichnung Flt-3 Ligand
Expressionssystem HEK293
Format Powder
Konjugat Unconjugated
Mehr Spezifikationen ansehen

Produkte 3
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16437549
Unterlagen ansehen
ACROBiosystems
FLL-H5223-1MG
1 mg
3224.10 CHF
1 Milligramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab
30-06-2021
Zum Warenkorb hinzufügen
 
15826777
Unterlagen ansehen
ACROBiosystems
FLL-H5223-20UG
20 µg
207.26 CHF
20 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab
30-06-2021
Zum Warenkorb hinzufügen
 
15836777
Unterlagen ansehen
ACROBiosystems
FLL-H5223-50UG
50 µg
403.01 CHF
50 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab
30-06-2021
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

FMS-like tyrosine kinase 3 ligand (Flt-3 Ligand) is also known as FL, Flt3L and FLT3LG, is an α-helical cytokine that promotes the differentiation of multiple hematopoietic cell lineages. FLT3LG is expressed as a noncovalentlylinked dimer by T cells and bone marrow and thymic fibroblasts. Each 36 kDa chain carries approximately 12 kDa of N- and O- linked carbohydrates. FLT3LG is structurally homologous to stem cell factor (SCF) and colony stimulating facor 1 (CSF-1). FLT3LG acts as a growth factor that increases the number of immune cells by activating the hematopoietic progenitors. It also induces the mobilization of the hematopoietic progenitors and stem cells in vivo which may help the system to kill cancer cells. FLT3LG induces the expansion of monocytes and immature dendritic cells as well as early B cell lineage differentiation. FLT3LG cooperates with IL2, IL6, IL7, and IL15 to induce NK cell development and with IL3, IL7 and IL11 to induce terminal B cell maturation. Animal studies also show FLT3LG to reduce the severity of experimentally induced allergic inflammation. FLT3LG is crucial for steady-state pDC and cDC development. A lack of FLT3L results in low levels of DCs.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.9 kDa. The protein migrates as 28-30 kDa and 31-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

Human Flt-3 Ligand, His Tag (FLL-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Pro 185 (Accession # P49771-1).
HEK293
Unconjugated
-20°C
Flt-3 Ligand
Powder
Cytokines and Growth Factors
Sicherheitsdatenblatt (SDS)