Beschreibung
Human Fibronectin Native Protein for Western Blot, IHC, ELISA, Ctrl, FN
Natural human fibronectin is intended for use in cell culture applications. Fibronectin is involved in a number of cellular processes including cell adhesion, migration, morphology, and regulation of cell growth and differentiation.
- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
Shipping Condition: Dry Ice.
Spezifikation
Spezifikation
| Zur Verwendung mit (Anwendung) | Cell Culture |
| Menge | 1 mg |
| Gen-Alias | Fibronectin Natural Human |
| Spezies | Human |
| Inhalt und Lagerung | Store in freezer (-5 to -30°C). |
| Versandbedingung | Dry Ice |
| Protein-Subtyp | Fibronectin |
| Forschungskategorie | Extracellular Matrix & Cell Adhesion, Stem Cell Research |
| Produktlinie | Gibco |