missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human FGFR4 Partial ORF (AAH11847.1, 31 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal Artikelnummer.: 16128114

Abnova™ Human FGFR4 Partial ORF (AAH11847.1, 31 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal

Artikelnummer. 16128114
25 ug, 25 Mikrogramm
Click to view available options
Menge:
10 µg
25 ug
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 16128114

Marke: Abnova™ H00002264Q01.25ug

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the fibroblast growth factor receptor family, where amino acid sequence is highly conserved between members and throughout evolution. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein would consist of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. The genomic organization of this gene, compared to members 1-3, encompasses 18 exons rather than 19 or 20. Although alternative splicing has been observed, there is no evidence that the C-terminal half of the IgIII domain of this protein varies between three alternate forms, as indicated for members 1-3. This particular family member preferentially binds acidic fibroblast growth factor and, although its specific function is unknown, it is overexpressed in gynecological tumor samples, suggesting a role in breast and ovarian tumorigenesis. [provided by RefSeq]

Sequence: EPCLAPSLEQQEQELTVALGQPVRLCCGRAERGGHWYKEGSRLAPAGRVRGWRGRLEIASFLPEDAGRYLCLARGSMIVLQNLTLITGDSLTSSNDDEDPKSHRDPSNRHSYPQQ

Spezifikation

Zugriffsnummer AAH11847.1
Zur Verwendung mit (Anwendung) Antibody Production, ELISA, Protein Array, Western Blot
Zusammensetzung 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gen-ID (Entrez) 2264
Molekulargewicht 38.39kDa
Name FGFR4 (Human) Recombinant Protein (Q01)
Qualitätskontrollen 12.5% SDS-PAGE Stained with Coomassie Blue.
Menge 25 ug
Immunogen EPCLAPSLEQQEQELTVALGQPVRLCCGRAERGGHWYKEGSRLAPAGRVRGWRGRLEIASFLPEDAGRYLCLARGSMIVLQNLTLITGDSLTSSNDDEDPKSHRDPSNRHSYPQQ
Lagerungsbedingungen Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Kennzeichnung RUO
Gen-Alias CD334/JTK2/MGC20292/TKF
Gebräuchliche Bezeichnung FGFR4
Gensymbol FGFR4
Spezies Wheat Germ (in vitro)
Rekombinant Recombinant
Proteinmarkierung GST
Expressionssystem wheat germ expression system
Form Liquid
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Abnova™ Human FGFR4 Partial ORF (AAH11847.1, 31 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt