missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FCRL5 Partial ORF (NP_112571.1, 105 a.a. - 214 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00083416-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: LILQAPLSVFEGDSVVLRCRAKAEVTLNNTIYKNDNVLAFLNKRTDFHIPHACLKDNGAYRCTGYKESCCPVSSNTVKIQVQEPFTRPVLRASSFQPISGNPVTLTCETQSpezifikation
NP_112571.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LILQAPLSVFEGDSVVLRCRAKAEVTLNNTIYKNDNVLAFLNKRTDFHIPHACLKDNGAYRCTGYKESCCPVSSNTVKIQVQEPFTRPVLRASSFQPISGNPVTLTCETQ | |
RUO | |
FCRL5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
83416 | |
FCRL5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BXMAS1/CD307/DKFZp667E2019/DKFZp667F216/FCRH5/FLJ00333/FLJ00397/IRTA2/MGC119590/MGC119592/MGC119593/PRO820 | |
FCRL5 | |
Recombinant | |
wheat germ expression system |