missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Fc gamma RIIB (CD32b) His-tag, Recombinant Protein
HEK293 cells Recombinant Protein
110.00 CHF - 346.00 CHF
Spezifikation
Zugriffsnummer | P31994 |
---|---|
Zur Verwendung mit (Anwendung) | Control, ELISA, Immunohistochemistry, Western Blot |
Zusammensetzung | PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 |
Gen-ID (Entrez) | 2213 |
Name | Human Fc gamma RIIB (CD32b) His-tag |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
17154193
|
Invitrogen™
A42519 |
5 μg |
110.00 CHF
5 Mikrogramm |
Verfügbar ab: 29-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
17164193
|
Invitrogen™
A42520 |
50 μg |
346.00 CHF
50 Mikrogramm |
Verfügbar ab: 29-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.Spezifikation
P31994 | |
PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human Fc gamma RIIB (CD32b) His-tag | |
-20°C | |
<1 EU/μg | |
FCGR2B | |
Unconjugated | |
Lyophilized |
Control, ELISA, Immunohistochemistry, Western Blot | |
2213 | |
HEK293 cells | |
RUO | |
AI528646; CD32; CD32 receptor 2; CD32 receptor 2 A; CD32 receptor 2 b; CD32A; CD32B; CDw32; F630109E10Rik; Fc fragment of IgG receptor IIb; Fc fragment of IgG, low affinity II, receptor for (CD32); Fc fragment of IgG, low affinity IIb, receptor; Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor for (CD32); Fc gamma receptor IIb; Fc gamma RIIb; Fc receptor, IgG, low affinity IIb; Fc[g]RII; FCG2; fc-gamma RII; fc-gamma RII-a; Fc-gamma RII-b; Fc-gamma-RII; fc-gamma-RIIa; FcgammaRIIB; fc-gamma-RIIb; FcgammaRIIB2; FcGR; FCGR2; Fcgr2a; FCGR2A1; FCGR2B; FcgRII; Fcr-2; Fcr-3; FCRII; fcRII-a; fcRII-b; FGFR2B; IGFR2; IgG Fc receptor II beta; igG Fc receptor II-b; Low affinity immunoglobulin gamma Fc region receptor II; low affinity immunoglobulin gamma Fc region receptor II-b; Ly-17; Lym-1; Ly-m20; Lymphocyte antigen 17 | |
Human | |
Recombinant | |
≥ 90% |