missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Fc gamma RIIA (CD32a) His-tag, Recombinant Protein
HEK293 cells Recombinant Protein
79.75 CHF - 247.00 CHF
Spezifikation
Zugriffsnummer | P12318 |
---|---|
Zur Verwendung mit (Anwendung) | Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot |
Zusammensetzung | PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 |
Gen-ID (Entrez) | 2212 |
Name | Human Fc gamma RIIA (CD32a) His-tag |
Produktcode | Marke | Menge | Name | Name | Preis | Menge & Verfügbarkeit | |||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Name | Name | Preis | Menge & Verfügbarkeit | |||
17124203
|
Invitrogen™
A42529 |
50 μg | Human Fc gamma RIIA (CD32a) His-tag |
247.00 CHF
50 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||
17114203
|
Invitrogen™
A42528 |
5 μg | Human Fc gamma RIIA (CD32a) His-tag |
79.75 CHF
5 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||
Beschreibung
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.Spezifikation
P12318 | |
PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human Fc gamma RIIA (CD32a) His-tag | |
-20°C | |
<1 EU/μg | |
FCGR2A | |
Unconjugated | |
Lyophilized |
Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot | |
2212 | |
HEK293 cells | |
RUO | |
CD16; CD32; CD32A; CDw32; Fc fragment of IgG low affinity IIa receptor for (CD32); Fc fragment of IgG receptor IIa; Fc fragment of IgG, low affinity IIa, receptor; Fc fragment of IgG, low affinity IIa, receptor (CD32); Fc gamma receptor IIa; Fc receptor, IgG, low affinity III; FCG2; fc-gamma RII-a; Fc-gamma RIII; fc-gamma-RIIa; FcGR; FCGR2; FCGR2A; FCGR2A1; Fcgr3; Fcgr3a; fcRII-a; fcRIII; FGCR; IGFR2; IgG Fc receptor II-a; igG Fc receptor III; Immunoglobulin G Fc receptor II; low affinity immunoglobulin gamma Fc region receptor II-a; Low affinity immunoglobulin gamma Fc region receptor III | |
Human | |
Recombinant | |
≥ 90% |