missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Fc gamma RI (CD64) His-tag, Recombinant Protein
CHO cells Recombinant Protein
Marke: Invitrogen™ A42517
Additional Details : Gewicht : 0.01060kg
Beschreibung
CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.Spezifikation
P12314 | |
PBS with 0.01% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human Fc gamma RI (CD64) His-tag | |
CHO cells | |
RUO | |
AI323638; AV092959; CD64; CD64 antigen; CD64A; Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG receptor Ia; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc receptor, IgG, high affinity I; FCG1; Fc-gamma receptor 1; Fc-gamma receptor I A1; fc-gamma RI; Fc-gamma RIA; FcgammaRI; FcgammaRIa; Fcgr1; FCGR1A; Fcgr1b; FcgRI; FCRI; FLJ18345; high affinity IgG Fc receptor, subunit beta; high affinity immunoglobulin gamma Fc receptor I; high-affinity immunoglobulin gamma Fc receptor I; IGFR1; IgG Fc receptor I; IgG high affinity Fc receptor; IGGHAFC; RP11-196G18.2 | |
Human | |
Recombinant | |
>95% |
Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot | |
2209 | |
5 μg | |
-20°C | |
<1 EU/μg | |
FCGR1A | |
Unconjugated | |
Lyophilized |