Learn More
Abnova™ Human FBXO3 Full-length ORF (AAH46110, 1 a.a. - 471 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternative splicing of this gene generates 2 transcript variants diverging at the 3' end. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | AAH46110 |
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 26273 |
Molekulargewicht | 77.55kDa |
Name | FBXO3 (Human) Recombinant Protein (P01) |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 10 μg |
Immunogen | MAAMETETAPLTLESLPTDPLLLILSFLDYRDLINCCYVSRRLSQLSSHDPLWRRHCKKYWLISEEEKTQKNQCWKSLFIDTYSDVGRYIDHYAAIKKAWDDLKKYLEPRCPRMVLSLKEGAREEDLDAVEAQIGCKLPDDYRCSYRIHNGQKLVVPGLLGSMALSNHYRSEDLLDVDTAAGGFQQRQGLKYCLPLTFCIHTGLSQYIAVEAAEGRNKNEVFYQCPDQMARNPAAIDMFIIGATFTDWFTSYVKNVVSGGFPIIRDQIFRYVHDPECVATTGDITVSVSTSFLPELSSVHPPHYFFTYRIRIEMSKDALPEKACQLDSRYWRITNAKGDVEEVQGPGVVGEFPIISPGRVYEYTSCTTFSTTSGYMEGYYTFHFLYFKDKIFNVAIPRFHMACPTFRVSIARLEMGPDEYEEMEEEEEEEEEEDEDDDSADMDESDEDDEEERRRRVFDVPIRRRRCSRLF |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.