Learn More
Abnova™ Human FBXO17 Partial ORF (NP_079183, 34 a.a. - 133 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_079183 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 115290 |
Molekulargewicht | 36.74kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16121477
|
Abnova™
H00115290-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 29-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16131477
|
Abnova™
H00115290-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 29-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it contains an F-box domain. Alternative splicing of this gene results in 2 transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: PPRSLVTRCRPVCRAWRDIVDGPTVWLLQLARDRSAEGRALYAVAQRCLPSNEDKEEFPLCALARYCLRAPFGRNLIFNSCGEQGFRGWEVEHGGNGWAISpezifikation
NP_079183 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBG4/FBX26/FBXO26/FLJ11798/FLJ25205/Fbx17/MGC9379 | |
FBXO17 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
115290 | |
FBXO17 (Human) Recombinant Protein (Q01) | |
PPRSLVTRCRPVCRAWRDIVDGPTVWLLQLARDRSAEGRALYAVAQRCLPSNEDKEEFPLCALARYCLRAPFGRNLIFNSCGEQGFRGWEVEHGGNGWAI | |
RUO | |
FBXO17 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |