missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FBXO10 Partial ORF (XP_291314, 863 a.a. - 962 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | XP_291314 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 26267 |
Molekulargewicht | 36.74kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16138252
|
Abnova™
H00026267-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16128252
|
Abnova™
H00026267-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Members of the F-box protein family, such as FBXO10, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]
Sequence: RAKALVQENIIFQGKTSKTIFQQISNNRECIMQNNKFLVFKKKSDTWRLVNPPARPHLENSLRRPSAAHNGQKVTAMATRITARVEGGYHSNRSVFCTILSpezifikation
XP_291314 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBX10/FLJ41992/MGC149840 | |
FBXO10 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
26267 | |
FBXO10 (Human) Recombinant Protein (Q01) | |
RAKALVQENIIFQGKTSKTIFQQISNNRECIMQNNKFLVFKKKSDTWRLVNPPARPHLENSLRRPSAAHNGQKVTAMATRITARVEGGYHSNRSVFCTIL | |
RUO | |
FBXO10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |