missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FAM9C Full-length ORF (NP_777561.1, 1 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00171484-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MAAKDQLEVQVMAAQEMELAGKDPVSHEHEERKPVTETKEGDVTDEHGERGSFAETDEHTGVDTKELEDIAADIKEHLAAKRKRIEKIAKACSEIKNRIKNVLRTTQLKRQKRDYRISLKLPNVLEEFITDEQKDEEGDGEKEEQIKIFQEQQKRWQQDGKGTERDSpezifikation
NP_777561.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FAM9C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
171484 | |
FAM9C (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAKDQLEVQVMAAQEMELAGKDPVSHEHEERKPVTETKEGDVTDEHGERGSFAETDEHTGVDTKELEDIAADIKEHLAAKRKRIEKIAKACSEIKNRIKNVLRTTQLKRQKRDYRISLKLPNVLEEFITDEQKDEEGDGEKEEQIKIFQEQQKRWQQDGKGTERD | |
RUO | |
FAM9C | |
Recombinant | |
wheat germ expression system |