missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FAM108A1 Full-length ORF (NP_112490.2, 1 a.a. - 361 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00081926-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MNGLSLSELCCLFCCPPCPGRIAAKLAFLPPEATYSLVPEPEPGPGGAGAAPLGTLRASSGAPGRWKLHLTERADFQYSQRELDTIEVFPTKSARGNRVSCMYVRCVPGARQGHQAQGGHPQLAWVGRLGDSNNPAPGGCLLGESWGTGAALACGYIHLLARYTVLFSHGNAVDLGQMSSFYIGLGSRLHCNIFSYDYSGYGASSGRPSERNLYADIDAAWQALRTRYGISPDSIILYGQSIGTVPTVDLASRYECAAVVLHSPLTSGMRVAFPDTKKTYCFDAFPNIEKVSKITSPVLIIHGTEDEVIDFSHGLALYERCPKAVEPLWVEGAGHNDIELYSQYLERLRRFISQELPSQRASpezifikation
NP_112490.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C19orf27/MGC5244 | |
FAM108A1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
81926 | |
FAM108A1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNGLSLSELCCLFCCPPCPGRIAAKLAFLPPEATYSLVPEPEPGPGGAGAAPLGTLRASSGAPGRWKLHLTERADFQYSQRELDTIEVFPTKSARGNRVSCMYVRCVPGARQGHQAQGGHPQLAWVGRLGDSNNPAPGGCLLGESWGTGAALACGYIHLLARYTVLFSHGNAVDLGQMSSFYIGLGSRLHCNIFSYDYSGYGASSGRPSERNLYADIDAAWQALRTRYGISPDSIILYGQSIGTVPTVDLASRYECAAVVLHSPLTSGMRVAFPDTKKTYCFDAFPNIEKVSKITSPVLIIHGTEDEVIDFSHGLALYERCPKAVEPLWVEGAGHNDIELYSQYLERLRRFISQELPSQRA | |
RUO | |
FAM108A1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |