Learn More
ACROBiosystems Human FABP8 / PMP2 Protein, His Tag
Marke: ACROBiosystems PM2-H5149-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Myelin P2 protein (PMP2) is also known as peripheral myelin protein 2, which is a cytosolic protein found primarily in peripheral nerves. PMP2 is a small, basic, and cytoplasmic lipid binding protein of peripheral myelin. Also, PMP2 may play a role in lipid transport protein in Schwann cells. Furthermore, PMP2 may bind cholesterol.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15.7 kDa. The protein migrates as 16 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
NP_002668.1 | |
1 mg | |
Ser 2 - Val 132 | |
PMP2,Myelin P2 protein,FABP8,M-FABP,MP2,P2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
15.7 kDa | |
Human FABP8, His Tag (PM2-H5149) is expressed from E.coli cells. It contains AA Ser 2 - Val 132 (Accession # P02689-1). | |
-20°C | |
FABP8 | |
Human | |
PBS, pH7.4 | |
E.coli | |
Other Recombinant Proteins |