Learn More
Abnova™ Human F11 native Protein from human plasma
Beschreibung
This gene encodes coagulation factor XI of the blood coagulation cascade. This protein is present in plasma as a zymogen, which is a unique plasma coagulation enzyme because it exists as a homodimer consisting of two identical polypeptide chains linked by disulfide bonds. During activation of the plasma factor XI, an internal peptide bond is cleaved by factor XIIa (or XII) in each of the two chains, resulting in activated factor XIa, a serine protease composed of two heavy and two light chains held together by disulfide bonds. This activated plasma factor XI triggers the middle phase of the intrisic pathway of blood coagulation by activating factor IX. Defects in this factor lead to Rosenthal syndrome, a blood coagulation abnormality. [provided by RefSeq]
Spezifikation
Spezifikation
| Konzentration | 0.37 mg/mL |
| Zur Verwendung mit (Anwendung) | SDS-PAGE |
| Zusammensetzung | In 4mM sodium acetate, 0.15 M NaCl, pH 5.3 |
| Gen-ID (Entrez) | 2160 |
| Molekulargewicht | 80kDa |
| Name | F11 (Human) Native Protein |
| Vorbereitungsmethode | Native protein purified from human plasma |
| Reinigungsverfahren | Multi-step chromatography |
| Menge | 100 μg |
| Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.