missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ezrin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Marke: R&D Systems MAB7239-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Ezrin Monoclonal specifically detects Ezrin in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry.
Spezifikation
| Ezrin | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2, VIL2MGC1584, Villin 2, villin 2 (ezrin), villin-2 | |
| E. coli-derived recombinant human Ezrin Lys438-Arg562 Accession # P15311 | |
| 25 μg | |
| Primary | |
| Detects human Ezrin in direct ELISAs and Western blots. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
| 2086A | |
| Western Blot 0.2 ug/mL, Immunohistochemistry 1-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 1-15 ug/mL | |
| P15311 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 7430 | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur