missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EXOSC3 Partial ORF (NP_057126.2, 176 a.a. - 274 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00051010-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: PEMVCIDSCGRANGMGVIGQDGLLFKVTLGLIRKLLAPDCEIIQEVGKLYPLEIVFGMNGRIWVKAKTIQQTLILANILEACEHMTSDQRKQIFSRLAESpezifikation
NP_057126.2 | |
Liquid | |
51010 | |
EXOSC3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CGI-102/MGC15120/MGC723/RP11-3J10.8/RRP40/Rrp40p/bA3J10.7/hRrp40p/p10 | |
EXOSC3 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PEMVCIDSCGRANGMGVIGQDGLLFKVTLGLIRKLLAPDCEIIQEVGKLYPLEIVFGMNGRIWVKAKTIQQTLILANILEACEHMTSDQRKQIFSRLAE | |
RUO | |
EXOSC3 | |
Wheat Germ (in vitro) | |
GST |