Learn More
R&D Systems™ Human EVC2 (NP_667338) VersaClone cDNA
Beschreibung
The Human EVC2 (NP_667338) VersaClone cDNA clone contains the complete ORF for EVC2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The EVC2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Spezifikation
Spezifikation
| Gen | hEVC2 |
| Zugriffsnummer | NP_667338 |
| Konzentration | 10 μg at 0.2 μg/μL |
| Zusammensetzung | cDNA is provided in 10 mM Tris-Cl |
| Einsatztyp | 3940bp |
| pH | 8.5 |
| Gen-ID (Entrez) | 132884 |
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Spezies | Human |
| Produktlinie | R&D Systems |
| Mehr anzeigen |