missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EVC Partial ORF (NP_055371, 493 a.a. - 602 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_055371 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 2121 |
Molekulargewicht | 37.84kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16127834
|
Abnova™
H00002121-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 31-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16117834
|
Abnova™
H00002121-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 31-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a protein containing a leucine zipper and a transmembrane domain. This gene has been implicated in both Ellis-van Creveld syndrome (EvC) and Weyers acrodental dysostosis. [provided by RefSeq]
Sequence: VLERQRLMQCDLEEEENVRATEAVVALCQELYFSTVDTFQKFVDALFLQTLPGMTGLPPEECDYLRQEVQENAAWQLGKSNRFRRQQWKLFQELLEQDQQVWMEECALSSSpezifikation
NP_055371 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DWF-1/EVC1/EVCL/MGC105107 | |
EVC | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
2121 | |
EVC (Human) Recombinant Protein (Q01) | |
VLERQRLMQCDLEEEENVRATEAVVALCQELYFSTVDTFQKFVDALFLQTLPGMTGLPPEECDYLRQEVQENAAWQLGKSNRFRRQQWKLFQELLEQDQQVWMEECALSS | |
RUO | |
EVC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |