missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Estradiol Rapid ELISA Kit

Artikelnummer. 30827015
Change view
Click to view available options
Menge:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30827015 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30827015 Lieferant Invitrogen™ Lieferanten-Nr. EELR003

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

ELISA

The Human Estradiol (E2) ELISA quantitates E2 in serum, plasma, urine, saliva. Principle of the method The Human E2 solid phase competitive ELISA (enzyme linked immunosorbent assay) is designed to measure the amount of the target bound between the pre coated antigen and the HRP linked antibody specific for Human E2. A target specific antigen has been pre coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells, where Human E2 in the samples (or standards) competes with the immobilized (capture) antigen for sites on the HRP linked detection antibody. Excess conjugate and unbound sample or standard are washed away from the plate. The substrate solution is added to each well, and the enzyme substrate reaction is terminated by the addition of a stop solution, turning the color yellow. The optical density (OD) is measured spectrophotometrically at a wavelength of 450±2 nm. The intensity of this signal is inversely proportional to the concentration of Human E2 present in the original specimen, which is determined by comparing the OD of the samples to the standard curve. Achieve fast and reliable results in just 90 minutes with our Rapid ELISA kit, featuring a streamlined protocol that requires only one wash step. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on v...

Estradiol (17 beta-estradiol) is a sex hormone. Estradiol is the predominant sex hormone present in females. It is also present in males, being produced as an active metabolic product of testosterone. It represents the major estrogen in humans. Estradiol has not only a critical impact on reproductive and sexual functioning, but also affects other organs including the bones.
TRUSTED_SUSTAINABILITY

Spezifikation

Assay-Bereich 31.25-2000 pg/mL
Assay-Empfindlichkeit: 13.22 pg/mL
Konjugat HRP
Immunassay-Kit-Format Coated ELISA Kit
Produkttyp ELISA
Probentyp Plasma, Saliva, Serum, Urine
Zielspezies Human
Zur Verwendung mit (Geräte) Absorbance Microplate Reader
Gen-Alias 17-Beta-Estradiol; 17-beta-HSD17-Beta-Estradiol; 17-beta-HSD;
Interassay-VK <10%
Intraassay-VK <10%
Kitinhalt Pre-coated 96 well plate, Standard, HRP Conjugate, Standard & Sample Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, HRP Conjugate, Standard & Sample Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Marker oder Farbstoff HRP
Menge 96 Tests
Kennzeichnung RUO
Probenvolumen Plasma 50 μL, Saliva 50 μL, Serum 50 μL, Urine 50 μL
Versandbedingung Wet or Dry Ice
Lagerungsbedingungen Refer to product documentation for component specific storage temperature.
Ziel Estradiol
Prüfzeit 15 min
Gesamte Assay-Zeit 1 hr 30 min
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.