missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Erythropoietin R Antibody, R&D Systems™

Mouse Monoclonal Antibody
264.00 CHF - 559.00 CHF
Spezifikation
| Antigen | Erythropoietin R |
|---|---|
| Klon | 1052708 |
| Verdünnung | Immunocytochemistry 3-25 ug/mL |
| Anwendungen | Immunocytochemistry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18770557
![]() |
R&D Systems
MAB11190-SP |
25 μg |
264.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18781427
![]() |
R&D Systems
MAB11190-100 |
100 μg |
559.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Erythropoietin R Monoclonal antibody specifically detects Erythropoietin R in Human samples. It is validated for ImmunocytochemistrySpezifikation
| Erythropoietin R | |
| Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| EpoR, EPO-R, erythropoietin receptor, MGC138358 | |
| Mouse myeloma cell line NS0-derived recombinant human Erythropoietin R, Ala25-Pro250, Accession # P19235 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1052708 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P19235 | |
| 2057 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts