missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ErbB2 (without Catalytic Activity Domain) Fc Chimera, Recombinant Protein
HEK293 cells Recombinant Protein
109.00 CHF - 247.00 CHF
Spezifikation
Zugriffsnummer | P04626 |
---|---|
Zur Verwendung mit (Anwendung) | Control, ELISA, Immunohistochemistry, Western Blot |
Zusammensetzung | protein with 0.02% Tween 80, 140 mM NaCl, 2.7 mM KCl, 5% mannitol, 5% trehalose and no preservative; pH 7.4 |
Gen-ID (Entrez) | 2064 |
Name | Human ErbB2 (without Catalytic Activity Domain) Fc Chimera |
Produktcode | Marke | Menge | Gen-Alias | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Gen-Alias | Preis | Menge & Verfügbarkeit | ||||
17104173
|
Invitrogen™
A42493 |
5 μg | avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; Proto-oncogene c-ErbB-2; proto-oncogene NEU; Receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb |
109.00 CHF
5 Mikrogramm |
Verfügbar ab: 29-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||
17114173
|
Invitrogen™
A42494 |
50 μg | avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; Proto-oncogene c-ErbB-2; proto-oncogene NEU; Receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb |
247.00 CHF
50 Mikrogramm |
Verfügbar ab: 29-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||
Beschreibung
Protein tyrosine kinase that is part of several cell surface receptor complexes, but that apparently needs a coreceptor for ligand binding. Essential component of a neuregulin-receptor complex, although neuregulins do not interact with it alone. GP30 is a potential ligand for this receptor. Regulates outgrowth and stabilization of peripheral microtubules (MTs). Upon ERBB2 activation, the MEMO1-RHOA-DIAPH1 signaling pathway elicits the phosphorylation and thus the inhibition of GSK3B at cell membrane. This prevents the phosphorylation of APC and CLASP2, allowing its association with the cell membrane. In turn, membrane-bound APC allows the localization of MACF1 to the cell membrane, which is required for microtubule capture and stabilization.Spezifikation
P04626 | |
protein with 0.02% Tween 80, 140 mM NaCl, 2.7 mM KCl, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human ErbB2 (without Catalytic Activity Domain) Fc Chimera | |
-20°C | |
<1 EU/μg | |
ERBB2 | |
Unconjugated | |
Lyophilized |
Control, ELISA, Immunohistochemistry, Western Blot | |
2064 | |
HEK293 cells | |
RUO | |
avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; Proto-oncogene c-ErbB-2; proto-oncogene NEU; Receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb | |
Human | |
Recombinant | |
≥ 85% |