Learn More
R&D Systems™ Human Embigin/EMB (NP_940851) VersaClone cDNA
Beschreibung
The Human Embigin/EMB (NP_940851) VersaClone cDNA clone contains the complete ORF for Embigin homolog, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Embigin homolog insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Spezifikation
Spezifikation
| Gen | hEMB |
| Zugriffsnummer | NP_940851 |
| Konzentration | 10 μg at 0.2 μg/μL |
| Zusammensetzung | cDNA is provided in 10 mM Tris-Cl |
| Einsatztyp | 997bp |
| pH | 8.5 |
| Gen-ID (Entrez) | 133418 |
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Spezies | Human |
| Produktlinie | R&D Systems |
| Mehr anzeigen |