missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DPPIV/CD26 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 27 publications
Marke: R&D Systems AF1180-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
DPPIV/CD26 Polyclonal specifically detects DPPIV/CD26 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
Spezifikation
| DPPIV/CD26 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPP4, DPPIV, EC 3.4.14.5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1803 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Simple Western 2 ug/mL, Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 1-15 ug/mL | |
| Q53TN1 | |
| DPP4 | |
| Mouse myeloma cell line NS0-derived recombinant human DPPIV/CD26 (Catalog # 1180-SE ) Asp34-Pro766 Accession # Q53TN1 | |
| 25 μg | |
| Primary | |
| Detects human DPPIV/CD26 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur