missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DNMBP Full-length ORF (AAH41628.1, 1 a.a. - 823 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00023268-P01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16127112

  • 379.00 CHF / 10 Mikrogramm
Verfügbar ab: 17-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

DNMBP belongs to the DBL (MIM 311030) family of guanine nucleotide exchange factors and plays a role in the regulation of cell junctions (Otani et al., 2006 [PubMed 17015620]).[supplied by OMIM]

Sequence: MTLLSSQSSSLVAPSGSVSAENPEQRMLEKRAKVIEELLQTERDYIRDLEMCIERIMVPMQQAQVPNIDFEGLFGNTQMVIKVSKQLLAALEISDAVGPVFLGHRDELEGTYKIYCQNHDEAIALLEIYEKDEKIQKHLQDSLADLKSLYNEWGCTNYINLGSFLIKPVQRVMRYPLLLMELLNSTPESHPDKVPLTNAVLAVKEINVNINEYKRRKDLVLKYRKGDEDSLMEKISKLNIHSIIKKSNRVSSHLKHLTGFAPQIKDEVFEETEKNFRMQERLIKSFIRDLSLYLQHIRESACVKVVAAVSMWDVCMERGHRDLEQFERVHRYISDQLFTNFKERTERLVISPLNQLLSMFTGPHKLVQKRFDKLLDFYNCTERAEKLKDKKTLEELQSARNNYEALNAQLLDELPKFHQYAQGLFTNCVHGYAEAHCDFVHQALEQLKPLLSLLKVAGREGNLIAIFHEEHSRVLQQLQVFTFFPESLPATKKPFERKTIDRQSARKPLLGLPSYMLQSEELRASLLARYPPEKLFQAERNFNAAQDLDVSLLEGDLVGVIKKKDPMGSQNRWLIDNGVTKGFVYSSFLKPYNPRRSHSDASVGSHSSTESEHGSSSPRFPRQNSGSTLTFNPSSMAVSFTSGSCQKQPQDASPPPKEWDQGTLSASLNPSNSESSPSRCPSDPDSTSQPRSGDSADVARDVKQPTATPRSYRNFRHPEIVGYSVPGRNGQSQDLVKGCARTAQAPEDRSTEPDGSEAEGNQVYFAVYTFKARNPNELSVSANQKLKILEFKDVTGNTEWWLAEVNGKKGYVPSNYIRKTEYT
Spezifikation

Spezifikation

AAH41628.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
120kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1010/TUBA
DNMBP
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
23268
DNMBP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTLLSSQSSSLVAPSGSVSAENPEQRMLEKRAKVIEELLQTERDYIRDLEMCIERIMVPMQQAQVPNIDFEGLFGNTQMVIKVSKQLLAALEISDAVGPVFLGHRDELEGTYKIYCQNHDEAIALLEIYEKDEKIQKHLQDSLADLKSLYNEWGCTNYINLGSFLIKPVQRVMRYPLLLMELLNSTPESHPDKVPLTNAVLAVKEINVNINEYKRRKDLVLKYRKGDEDSLMEKISKLNIHSIIKKSNRVSSHLKHLTGFAPQIKDEVFEETEKNFRMQERLIKSFIRDLSLYLQHIRESACVKVVAAVSMWDVCMERGHRDLEQFERVHRYISDQLFTNFKERTERLVISPLNQLLSMFTGPHKLVQKRFDKLLDFYNCTERAEKLKDKKTLEELQSARNNYEALNAQLLDELPKFHQYAQGLFTNCVHGYAEAHCDFVHQALEQLKPLLSLLKVAGREGNLIAIFHEEHSRVLQQLQVFTFFPESLPATKKPFERKTIDRQSARKPLLGLPSYMLQSEELRASLLARYPPEKLFQAERNFNAAQDLDVSLLEGDLVGVIKKKDPMGSQNRWLIDNGVTKGFVYSSFLKPYNPRRSHSDASVGSHSSTESEHGSSSPRFPRQNSGSTLTFNPSSMAVSFTSGSCQKQPQDASPPPKEWDQGTLSASLNPSNSESSPSRCPSDPDSTSQPRSGDSADVARDVKQPTATPRSYRNFRHPEIVGYSVPGRNGQSQDLVKGCARTAQAPEDRSTEPDGSEAEGNQVYFAVYTFKARNPNELSVSANQKLKILEFKDVTGNTEWWLAEVNGKKGYVPSNYIRKTEYT
RUO
DNMBP
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote