missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human DNAM-1 / CD226 Protein, His Tag
315.25 CHF - 2105.87 CHF
Spezifikation
Zugriffsnummer | NP_006557 |
---|---|
Molekulargewicht | 27.9 kDa |
Quelle | Human DNAM-1, His Tag (DN1-H52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Accession # NP_006557). |
Immunogen | Glu 19 - Asn 247 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15816587
|
ACROBiosystems
DN1-H52H6-100UG |
100 μg |
315.25 CHF
100 Mikrogramm |
Verfügbar ab: 30-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15826587
|
ACROBiosystems
DN1-H52H6-1MG |
1 mg |
2105.87 CHF
1 Milligramm |
Verfügbar ab: 30-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
DNAX accessory molecule 1 (DNAM-1), a single-pass type I membrane protein, is also known as CD226 antigen and platelet and T cell activation antigen 1 (PTA1), which contains 2 Ig-like C2-type (immunoglobulin-like) domains. DNAM-1 is a ∼65 kDa glycoprotein expressed on the surface of natural killer cells, platelets, monocytes and a subset of T cells. DNAM-1 mediates cellular adhesion to other cells bearing its ligands, CD112 and CD155, and cross-linking DNAM-1 with antibodies causes cellular activation. Furthermore, DNAM-1 can interact with PVR and PVRL2.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.9 kDa. The protein migrates as 36-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_006557 | |
Human DNAM-1, His Tag (DN1-H52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Accession # NP_006557). | |
-20°C | |
DNAM-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
27.9 kDa | |
Glu 19 - Asn 247 | |
DNAM1,CD226,PTA1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |