missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DCAMKL3 Partial ORF (XP_047355, 1031 a.a. - 1140 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00085443-Q01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16151027

  • 302.00 CHF / 10 Mikrogramm


Verfügbar ab
16-01-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: EKLVRTRSCRRSPEANPASGEEGWKGDSHRSSPRNPTQELRRPSKSMDKKEDRGPEDQESHAQGAAKAKKDLVEVLPVTEEGLREVKKDTRPMSRSKHGGWLLREHQAGF
Spezifikation

Spezifikation

XP_047355
Liquid
85443
DCAMKL3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DCAMKL3|DCDC3C|KIAA1765
DCLK3
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EKLVRTRSCRRSPEANPASGEEGWKGDSHRSSPRNPTQELRRPSKSMDKKEDRGPEDQESHAQGAAKAKKDLVEVLPVTEEGLREVKKDTRPMSRSKHGGWLLREHQAGF
RUO
DCLK3
wheat germ expression system
Yes
Videos
Dokumentation