missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DCAMKL3 Partial ORF (XP_047355, 1031 a.a. - 1140 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Specifications
Zugriffsnummer | XP_047355 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 85443 |
Molekulargewicht | 37.84kDa |
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
16161027
|
Abnova™
H00085443-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16151027
|
Abnova™
H00085443-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Description
Sequence: EKLVRTRSCRRSPEANPASGEEGWKGDSHRSSPRNPTQELRRPSKSMDKKEDRGPEDQESHAQGAAKAKKDLVEVLPVTEEGLREVKKDTRPMSRSKHGGWLLREHQAGFSpecifications
XP_047355 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DCAMKL3/DCDC3C/KIAA1765 | |
DCLK3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
85443 | |
DCAMKL3 (Human) Recombinant Protein (Q01) | |
EKLVRTRSCRRSPEANPASGEEGWKGDSHRSSPRNPTQELRRPSKSMDKKEDRGPEDQESHAQGAAKAKKDLVEVLPVTEEGLREVKKDTRPMSRSKHGGWLLREHQAGF | |
RUO | |
DCLK3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |