Learn More
R&D Systems™ Human DAP10/HCST (NP_055081) VersaClone cDNA
Artikelnummer. 18732308
Ansicht ändern
Click to view available options
Menge:
10 μg
Packungsgröße:
10 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18732308 10 μg 10 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18732308 Lieferant R&D Systems™ Lieferanten-Nr. RDC2544
  • ONLINE EXCLUSIVE PRICE
    345.00 CHF
    325.50 CHF / 10 Mikrogramm
    Sparen 19.50 CHF
    5% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 26-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.
  • The Human DAP10/HCST (NP_055081) VersaClone cDNA clone contains the complete ORF for DAP10/HCST, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The DAP10/HCST insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY
Spezifikation
Gen hDAP10/HCST
Zugriffsnummer NP_055081
Konzentration 10 μg at 0.2 μg/μL
Zusammensetzung cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gen-ID (Entrez) 10870
Inhalt und Lagerung Store the unopened product at -20 to -80°C
Spezies Human
Menge 10 μg