missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human Cystatin S / CST4 Protein, His Tag

Marke:  ACROBiosystems CS4-H5223-1MG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15826487

  • 1612.06 CHF / 1 Milligramm


Verfügbar ab
04-06-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Cystatin-4 (CST4) is also known as Cystatin-S, Salivary acidic protein 1, which is a secreted protein, which belongs to the cystatin family. Cystatin-S is expressed in submandibular and sublingual saliva but not in parotid saliva (at protein level) and is also expressed in saliva, tears, urine and seminal fluid. Cystatin-4 / Cystatin S strongly inhibits papain and ficin, partially inhibits stem bromelain and bovine cathepsin C, but does not inhibit porcine cathepsin B or clostripain. Papain is inhibited non-competitively.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.0 kDa. The protein migrates as 15 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation

Spezifikation

AAH65714
HEK293
Human
15.0 kDa
1 mg
Ser 21 - Ala 141
CST4,Cystatin-4,Cystatin-S,Cystatin-S5,Cystatin-SA-III
Unconjugated
25 mM MES, 150 mM NaCl, pH6.0
Less than 1.0 EU / µg by the LAL method.
Other Recombinant Proteins
Powder
>95% as determined by SDS-PAGE
Human Cystatin S, His Tag (CS4-H5223) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ala 141 (Accession # AAH65714).
-20°C
Cystatin S
His Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation