missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CYB5R4 Partial ORF (NP_057314, 388 a.a. - 487 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00051167-Q01.10ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
NCB5OR is a flavohemoprotein that contains functional domains found in both cytochrome b5 (CYB5A; MIM 250790) and CYB5 reductase (see MIM 250800).[supplied by OMIM]
Sequence: VKLMFFNKTEDDIIWRSQLEKLAFKDKRLDVEFVLSAPISEWNGKQGHISPALLSEFLKRNLDKSKVLVCICGPVPFTEQGVRLLHDLNFSKNEIHSFTASpezifikation
NP_057314 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VKLMFFNKTEDDIIWRSQLEKLAFKDKRLDVEFVLSAPISEWNGKQGHISPALLSEFLKRNLDKSKVLVCICGPVPFTEQGVRLLHDLNFSKNEIHSFTA | |
RUO | |
CYB5R4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51167 | |
CYB5R4 (Human) Recombinant Protein (Q01) | |
10 μg | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NCB5OR/RP4-676J13.1/dJ676J13.1 | |
CYB5R4 | |
Recombinant | |
wheat germ expression system |