missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human CXCR2 Full-length ORF (NP_001548.1, 1 a.a. - 360 a.a.) Recombinant Protein with GST-tag at N-terminal Artikelnummer.: 16129307
Nur noch undefined auf Lager
Zum Warenkorb hinzufügen
compliance-icons

Abnova™ Human CXCR2 Full-length ORF (NP_001548.1, 1 a.a. - 360 a.a.) Recombinant Protein with GST-tag at N-terminal

Artikelnummer. 16129307
10 ug, 10 Mikrogramm
Click to view available options
Menge:
10 ug
25 ug
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 16129307

Marke: Abnova™ H00003579P01.10ug

Nur noch undefined auf Lager
Zum Warenkorb hinzufügen
Nur noch undefined auf Lager
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the G-protein-coupled receptor family. This protein is a receptor for interleukin 8 (IL8). It binds to IL8 with high affinity, and transduces the signal through a G-protein activated second messenger system. This receptor also binds to chemokine (C-X-C motif) ligand 1 (CXCL1/MGSA), a protein with melanoma growth stimulating activity, and has been shown to be a major component required for serum-dependent melanoma cell growth. This receptor mediates neutrophil migration to sites of inflammation. The angiogenic effects of IL8 in intestinal microvascular endothelial cells are found to be mediated by this receptor. Knockout studies in mice suggested that this receptor controls the positioning of oligodendrocyte precursors in developing spinal cord by arresting their migration. This gene, IL8RA, a gene encoding another high affinity IL8 receptor, as well as IL8RBP, a pseudogene of IL8RB, form a gene cluster in a region mapped to chromosome 2q33-q36. [provided by RefSeq]

Sequence: MEDFNMESDSFEDFWKGEDLSNYSYSSTLPPFLLDAAPCEPESLEINKYFVVIIYALVFLLSLLGNSLVMLVILYSRVGRSVTDVYLLNLALADLLFALTLPIWAASKVNGWIFGTFLCKVVSLLKEVNFYSGILLLACISVDRYLAIVHATRTLTQKRYLVKFICLSIWGLSLLLALPVLLFRRTVYSSNVSPACYEDMGNNTANWRMLLRILPQSFGFIVPLLIMLFCYGFTLRTLFKAHMGQKHRAMRVIFAVVLIFLLCWLPYNLVLLADTLMRTQVIQETCERRNHIDRALDATEILGILHSCLNPLIYAFIGQKFRHGLLKILAIHGLISKDSLPKDSRPSFVGSSSGHTSTTL

Spezifikation

Zugriffsnummer NP_001548.1
Zur Verwendung mit (Anwendung) Antibody Production, Protein Array, ELISA, Western Blot
Zusammensetzung 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gen-ID (Entrez) 3579
Molekulargewicht 67.2kDa
Name CXCR2 (Human) Recombinant Protein (P01)
Reinigungsverfahren Glutathione Sepharose 4 Fast Flow
Qualitätskontrollen 12.5% SDS-PAGE Stained with Coomassie Blue.
Menge 10 ug
Immunogen MEDFNMESDSFEDFWKGEDLSNYSYSSTLPPFLLDAAPCEPESLEINKYFVVIIYALVFLLSLLGNSLVMLVILYSRVGRSVTDVYLLNLALADLLFALTLPIWAASKVNGWIFGTFLCKVVSLLKEVNFYSGILLLACISVDRYLAIVHATRTLTQKRYLVKFICLSIWGLSLLLALPVLLFRRTVYSSNVSPACYEDMGNNTANWRMLLRILPQSFGFIVPLLIMLFCYGFTLRTLFKAHMGQKHRAMRVIFAVVLIFLLCWLPYNLVLLADTLMRTQVIQETCERRNHIDRALDATEILGILHSCLNPLIYAFIGQKFRHGLLKILAIHGLISKDSLPKDSRPSFVGSSSGHTSTTL
Lagerungsbedingungen Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Kennzeichnung RUO
Gen-Alias CD182/CDw128b/CMKAR2/IL8R2/IL8RA/IL8RB
Gebräuchliche Bezeichnung CXCR2
Gensymbol CXCR2
Spezies Wheat Germ (in vitro)
Rekombinant Recombinant
Proteinmarkierung GST
Expressionssystem wheat germ expression system
Form Liquid
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • CXCR2 (Human) Recombinant Protein (P01)
Signalwort
  • Achtung
Gefahrenkategorie
  • Akute Toxizität Kategorie 4
  • Schwere Augenschädigung/Augenreizung Kategorie 2
  • Hautreizende/-ätzende Wirkung Kategorie 2
Gefahrenhinweis
  • H302-Gesundheitsschädlich bei Verschlucken.
  • H315-Verursacht Hautreizungen.
  • H319-Verursacht schwere Augenreizung.
Sicherheitshinweis
  • P102-Darf nicht in die Hände von Kindern gelangen.
  • P103-Vor Gebrauch Kennzeichnungsetikett lesen.
  • P233-Behälter dicht verschlossen halten.
  • P264-Nach Gebrauch gründlich waschen.
  • P270-Bei Gebrauch nicht essen, trinken oder rauchen.
  • P280-Schutzhandschuhe/Schutzkleidung/Augenschutz/Gesichtsschutz tragen.
  • P301+P310-BEI VERSCHLUCKEN: Sofort GIFTINFORMATIONSZENTRUM/Arzt anrufen/
  • P303+P361+P353-BEI BERÜHRUNG MIT DER HAUT (oder dem Haar): Alle kontaminierten Kleidungsstücke sofort ausziehen. Haut mit Wasser abwaschen/duschen.
  • P305+P351+P338-BEI KONTAKT MIT DEN AUGEN: Einige Minuten lang behutsam mit Wasser spülen. Vorhandene Kontaktlinsen nach Möglichkeit entfernen. Weiter spülen.
  • P404-In einem geschlossenen Behälter aufbewahren.
  • P501b-Inhalt/Behälter gemäß den örtlichen/regionalen/nationalen/internationalen Vorschriften zuführen.
Ergänzende Informationen
  • MIXTURE LIST-Enthält: tris-HCl, reduced glutathione
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Abnova™ Human CXCR2 Full-length ORF (NP_001548.1, 1 a.a. - 360 a.a.) Recombinant Protein with GST-tag at N-terminal >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt