missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CTF1 Full-length ORF (NP_001321.1, 1 a.a. - 201 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00001489-P01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16126787

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 09-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The protein encoded by this gene is a secreted cytokine that induces cardiac myocyte hypertrophy in vitro. It has been shown to bind and activate the ILST/gp130 receoptor. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MSRREGSLEDPQTDSSVSLLPHLEAKIRQTHSLAHLLTKYAEQLLQEYVQLQGDPFGLPSFSPPRLPVAGLSAPAPSHAGLPVHERLRLDAAALAALPPLLDAVCRRQAELNPRAPRLLRRLEDAARQARALGAAVEALLAALGAANRGPRAEPPAATASAASATGVFPAKVLGLRVCGLYREWLSRTEGDLGQLLPGGSA
Spezifikation

Spezifikation

NP_001321.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
47.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CT-1/CT1
CTF1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
1489
CTF1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MSRREGSLEDPQTDSSVSLLPHLEAKIRQTHSLAHLLTKYAEQLLQEYVQLQGDPFGLPSFSPPRLPVAGLSAPAPSHAGLPVHERLRLDAAALAALPPLLDAVCRRQAELNPRAPRLLRRLEDAARQARALGAAVEALLAALGAANRGPRAEPPAATASAASATGVFPAKVLGLRVCGLYREWLSRTEGDLGQLLPGGSA
RUO
CTF1
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote