missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human COPZ2 Full-length ORF (NP_057513.1, 1 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_057513.1 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 51226 |
Molekulargewicht | 49.9kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16129792
|
Abnova™
H00051226-P01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 29-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16119792
|
Abnova™
H00051226-P01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 29-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a subunit of the coatomer protein complex, a seven-subunit complex that functions in the formation of COPI-type, non-clathrin-coated vesicles. COPI vesicles function in the retrograde Golgi-to-ER transport of dilysine-tagged proteins. This gene is similar to a related family member, and the two encoded proteins form distinct isotypes of the coatomer protein complex. [provided by RefSeq]
Sequence: MQRPEAWPRPHPGEGAAAAQAGGPAPPARAGEPSGLRLQEPSLYTIKAVFILDNDGRRLLAKYYDDTFPSMKEQMVFEKNVFNKTSRTESEIAFFGGMTIVYKNSIDLFLYVVGSSYENELMLMSVLTCLFESLNHMLRKNVEKRWLLENMDGAFLVLDEIVDGGVILESDPQQVIQKVNFRADDGGLTEQSVAQVLQSAKEQIKWSLLKSpezifikation
NP_057513.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MQRPEAWPRPHPGEGAAAAQAGGPAPPARAGEPSGLRLQEPSLYTIKAVFILDNDGRRLLAKYYDDTFPSMKEQMVFEKNVFNKTSRTESEIAFFGGMTIVYKNSIDLFLYVVGSSYENELMLMSVLTCLFESLNHMLRKNVEKRWLLENMDGAFLVLDEIVDGGVILESDPQQVIQKVNFRADDGGLTEQSVAQVLQSAKEQIKWSLLK | |
RUO | |
COPZ2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51226 | |
COPZ2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC23008 | |
COPZ2 | |
Recombinant | |
wheat germ expression system |