missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLNK Full-length ORF (BAG52635.1, 1 a.a. - 428 a.a.) Recombinant Protein with GST-tag at N-terminal

Recombinant proteins expressed from in vitro wheat germ system.

Marke:  Abnova™ H00116449-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16127343

  • 574.00 CHF / 25 Mikrogramm
Verfügbar ab: 09-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

MIST is a member of the SLP76 family of adaptors (see LCP2, MIM 601603; BLNK, MIM 604515). MIST plays a role in the regulation of immunoreceptor signaling, including PLC-gamma (PLCG1; MIM 172420)-mediated B cell antigen receptor (BCR) signaling and FC-epsilon R1 (see FCER1A, MIM 147140)-mediated mast cell degranulation (Cao et al., 1999 [PubMed 10562326]; Goitsuka et al., 2000, 2001 [PubMed 10744659] [PubMed 11463797]).[supplied by OMIM]

  • Good Folding
  • Excellent amino acids labeling

Immunogen Sequence: MNRQGNRKTTKEGSNDLKFQNFSLPKNRSWPRINSATGQYQRMNKPLLDWERNFAAVLDGAKGHSDDDYDDPELRMEETWQSIKILPARPIKESEYADTHYFKVAMDTPLPLDTRTSISIGQPTWNTQTRLERVDKPISKDVRSQN

Spezifikation

Spezifikation

BAG52635.1
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer.
CLNK (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MNRQGNRKTTKEGSNDLKFQNFSLPKNRSWPRINSATGQYQRMNKPLLDWERNFAAVLDGAKGHSDDDYDDPELRMEETWQSIKILPARPIKESEYADTHYFKVAMDTPLPLDTRTSISIGQPTWNTQTRLERVDKPISKDVRSQNIKGDASVRKNKIPLPPPRPLITLPKKYQPLPPEPESSRPPLSQRHTFPEVQRMPSQISLRDLSEVLEAEKVPHNQRKPESTHLLENQNTQEIPLAISSSSFTTSNHSVQNRDHRGGMQPCSPQRCQPPASCSPHENILPYKYTSWRPPFPKRSDRKDVQHNEWYIGEYSRQAVEEAFMKENKDGSFLVRDCSTKSKEEPYVLAVFYENKVYNVKIRFLERNQQFALGTGLRGDEKFDSVEDIIEHYKNFPIILIDGKDKTGVHRKQCHLTQPLPLTRHLLPL
RUO
CLNK
GST
Antibody Production, Protein Array, ELISA, Western Blot
116449
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC35197/MIST
CLNK
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only