Learn More
Abnova™ Human CLDN11 Full-length ORF (NP_005593.2, 1 a.a. - 207 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
auf Anfrage
Spezifikation
Name | CLDN11 (Human) Recombinant Protein (P01) |
---|---|
Rekombinant | Yes |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Molekulargewicht | 48.4kDa |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Beschreibung
The protein encoded by this gene belongs to the claudin family of tight junction associated proteins and is a major component of central nervous system myelin that is necessary for normal CNS function. There is growing evidence that the protein determines the permeability between layers of myelin sheaths via focal adhesion and, with its expression highly regulated during development, may play an important role in cellular proliferation and migration. In addition, the protein is a candidate autoantigen in the development of autoimmune demyelinating disease. [provided by RefSeq]
Sequence: MVATCLQVVGFVTSFVGWIGVIVTTSTNDWVVTCGYTIPTCRKLDELGSKGLWADCVMATGLYHCKPLVDILILPGYVQACRALMIAASVLGLPAILLLLTVLPCIRMGQEPGVAKYRRAQLAGVLLILLALCALVATIWFPVCAHRETTIVSFGYSLYAGWIGAVLCLVGGCVILCCAGDAQAFGENRFYYTAGSSSPTHAKSAHVSpezifikation
CLDN11 (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
48.4kDa |