missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLDN11 Full-length ORF (NP_005593.2, 1 a.a. - 207 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00005010-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16150978

  • 448.00 CHF / 25 Mikrogramm


Verfügbar ab
01-07-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: MVATCLQVVGFVTSFVGWIGVIVTTSTNDWVVTCGYTIPTCRKLDELGSKGLWADCVMATGLYHCKPLVDILILPGYVQACRALMIAASVLGLPAILLLLTVLPCIRMGQEPGVAKYRRAQLAGVLLILLALCALVATIWFPVCAHRETTIVSFGYSLYAGWIGAVLCLVGGCVILCCAGDAQAFGENRFYYTAGSSSPTHAKSAHV
Spezifikation

Spezifikation

NP_005593.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OSP|OTM
CLDN11
Yes
wheat germ expression system
Liquid
5010
CLDN11 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVATCLQVVGFVTSFVGWIGVIVTTSTNDWVVTCGYTIPTCRKLDELGSKGLWADCVMATGLYHCKPLVDILILPGYVQACRALMIAASVLGLPAILLLLTVLPCIRMGQEPGVAKYRRAQLAGVLLILLALCALVATIWFPVCAHRETTIVSFGYSLYAGWIGAVLCLVGGCVILCCAGDAQAFGENRFYYTAGSSSPTHAKSAHV
RUO
CLDN11
Wheat Germ (in vitro)
GST
Videos
Dokumentation