Learn More
Abnova™ Human CHST3 Partial ORF (NP_004264, 312 a.a. - 411 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00009469-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes an enzyme which catalyzes the sulfation of chondroitin, a proteoglycan found in the extracellular matrix and most cells which is involved in cell migration and differentiation. Mutations in this gene are associated with spondylepiphyseal dysplasia and humerospinal dysostosis. [provided by RefSeq]
Sequence: VAFAGKYKTWKKWLDDEGQDGLREEEVQRLRGNCESIRLSAELGLRQPAWLRGRYMLVRYEDVARGPLQKAREMYRFAGIPLTPQVEDWIQKNTQAAHDGSpezifikation
NP_004264 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VAFAGKYKTWKKWLDDEGQDGLREEEVQRLRGNCESIRLSAELGLRQPAWLRGRYMLVRYEDVARGPLQKAREMYRFAGIPLTPQVEDWIQKNTQAAHDG | |
RUO | |
CHST3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9469 | |
CHST3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C6ST/C6ST1/HSD | |
CHST3 | |
Recombinant | |
wheat germ expression system |