Learn More
Abnova™ Human CHRM3 Partial ORF (NP_000731, 1 a.a. - 67 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_000731 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 1131 |
Molekulargewicht | 33.11kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16143201
|
Abnova™
H00001131-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 31-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16133201
|
Abnova™
H00001131-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 31-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 3 controls smooth muscle contraction and its stimulation causes secretion of glandular tissue. [provided by RefSeq]
Sequence: MTLHNNSTTSPLFPNISSSWIHSPSDAGLPPGTVTHFGSYNVSRAAGNFSSPDGTTDDPLGGHTVWQSpezifikation
NP_000731 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.11kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HM3 | |
CHRM3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1131 | |
CHRM3 (Human) Recombinant Protein (Q01) | |
MTLHNNSTTSPLFPNISSSWIHSPSDAGLPPGTVTHFGSYNVSRAAGNFSSPDGTTDDPLGGHTVWQ | |
RUO | |
CHRM3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |