missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chorionic Gonadotropin alpha/beta (HCG) Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Marke: R&D Systems MAB97701-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
hCG Monoclonal specifically detects hCG in Human samples. It is validated for ELISA.
Spezifikation
| hCG | |
| Monoclonal | |
| Unconjugated | |
| P01233 and P01215 | |
| Chinese Hamster Ovary cell line, CHO-derived human Chorionic Gonadotropin alpha/beta (HCG) and Human CG alpha , Ser21-Gln165 and Ala25-Ser116, Accession # P01233 and P01215 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1177B | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur