missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CENTG2 Partial ORF (NP_055729.2, 672 a.a. - 781 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_055729.2 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 116987 |
Molekulargewicht | 37.84kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16141547
|
Abnova™
H00116987-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16151547
|
Abnova™
H00116987-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
CENTG2 belongs to an ADP-ribosylation factor GTPase-activating (ARF-GAP) protein family involved in membrane traffic and actin cytoskeleton dynamics (Nie et al., 2002 [PubMed 12388557]).[supplied by OMIM]
Sequence: LPCTELSLGQHLLRATADEDLRTAILLLAHGSRDEVNETCGEGDGRTALHLACRKGNVVLAQLLIWYGVDVTARDAHGNTALAYARQASSQECIDVLLQYGCPDERFVLMSpezifikation
NP_055729.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CENTG2/GGAP1/KIAA1099/MGC71657 | |
AGAP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
116987 | |
CENTG2 (Human) Recombinant Protein (Q01) | |
LPCTELSLGQHLLRATADEDLRTAILLLAHGSRDEVNETCGEGDGRTALHLACRKGNVVLAQLLIWYGVDVTARDAHGNTALAYARQASSQECIDVLLQYGCPDERFVLM | |
RUO | |
AGAP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |