Learn More
Abnova™ Human CDKL3 Partial ORF (AAH41799, 483 a.a. - 592 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
The protein encoded by this gene is a member of cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This gene was identified as a gene absent in leukemic patients with chromosome 5q deletion. This loss may be an important determinant of dysmyelopoiesis. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | AAH41799 |
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 51265 |
Molekulargewicht | 37.51kDa |
Name | CDKL3 (Human) Recombinant Protein (Q01) |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 25 μg |
Immunogen | GPIQSQMEKGIFNERTGHSDQMANENKRKLNFSRSDRKEFHFPELPVTIQSKDTKGMEVKQIKMLKRESKKTESSKIPTLLNVDQNQEKQEGGDGHCEGKNLKRNRFFFW |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.