Learn More
ACROBiosystems Human CD83 Protein, His Tag
Marke: ACROBiosystems CD3-H5223-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
CD83 antigen is also known as B-cell activation protein and Cell surface protein HB15. CD83 is a single-pass type I membrane protein which contains one Ig-like V-type (immunoglobulin-like) domain. CD83 is expressed by activated lymphocytes, Langerhans cells and interdigitating reticulum cells. CD83 may play a significant role in antigen presentation or the cellular interactions that follow lymphocyte activation.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.9 kDa. The protein migrates as 18 kDa and 22 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.Spezifikation
AAH30830.1 | |
1 mg | |
Thr 20 - Ala 143 | |
CD83,HB15,hCD83,BL11 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
14.9 kDa | |
Human CD83, His Tag (CD3-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Ala 143 (Accession # Q01151-1). | |
-20°C | |
CD83 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |